Learn More
Medchemexpress LLC Naphthol AS-BI | 1237-75-8 | 99.9% | 1 G
SDP
Catalog No. 5000410832
Encompass_Preferred
Naphthol AS-BI is a substrate of β-glucuronidase. When used for staining biological tissues, it produces a bright red effect.
  • Substrate of β-glucuronidase.
  • Produces a bright red effect after staining biological tissues.
  • For research use only.
Catalog No. 50-004-10832 Supplier Medchemexpress LLC Supplier No. HYD01641G
Naphthol AS-BI is a substrate of β-glucuronidase. When used for staining biological tissues, it produces a bright red effect.
  • Substrate of β-glucuronidase.
  • Produces a bright red effect after staining biological tissues.
  • For research use only.