Learn More
Medchemexpress LLC Naphthol AS-BI | 1237-75-8 | 99.9% | 25 MG
SDP
Catalog No. 5000412044
Encompass_Preferred
Naphthol AS-BI is a substrate of β-glucuronidase and produces a bright red effect after staining biological tissues. This fluorescent dye is suitable for various laboratory applications.
  • Produces a bright red effect after staining biological tissues.
  • Functions as a fluorescent dye.
Catalog No. 50-004-12044 Supplier Medchemexpress LLC Supplier No. 25MGHYD0164
Naphthol AS-BI is a substrate of β-glucuronidase and produces a bright red effect after staining biological tissues. This fluorescent dye is suitable for various laboratory applications.
  • Produces a bright red effect after staining biological tissues.
  • Functions as a fluorescent dye.