Learn More
Medchemexpress LLC Naphthol AS-BI | 1237-75-8 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYD01645MG
Encompass_Preferred
Naphthol AS-BI is a substrate of β-glucuronidase, producing a bright red effect after staining biological tissues.
  • Functions as a substrate for β-glucuronidase
  • Generates a bright red effect when staining biological tissues
Catalog No. 50-004-10318