Learn More
Medchemexpress LLC Naphthol AS-BI | 1237-75-8 | MFCD00004077 | 99.7% | 372.21 g·mol⁻1 | C18H14BrNO3 | 10 MG
SDP
Catalog No. 5000297662
Encompass_Preferred
Naphthol AS-BI is a chromogenic substrate for β-glucuronidase that produces a bright red color when reacted in biological tissues. It is used in histochemical staining and biochemical assays to visualize enzyme activity in research settings.
  • Chromogenic β-glucuronidase substrate producing bright red stain.
  • High reported purity (≈99.7%).
  • Molecular formula C18H14BrNO3 and molecular weight 372.21 g·mol⁻1.
  • Supplied as a 10 mg vial for laboratory assays.
  • Intended for research use only.
Catalog No. 50-002-97662 Supplier Medchemexpress LLC Supplier No. HYD016410MG
Naphthol AS-BI is a chromogenic substrate for β-glucuronidase that produces a bright red color when reacted in biological tissues. It is used in histochemical staining and biochemical assays to visualize enzyme activity in research settings.
  • Chromogenic β-glucuronidase substrate producing bright red stain.
  • High reported purity (≈99.7%).
  • Molecular formula C18H14BrNO3 and molecular weight 372.21 g·mol⁻1.
  • Supplied as a 10 mg vial for laboratory assays.
  • Intended for research use only.