Learn More
AdipoGen Naphthol AS-BI
SDP
Catalog No. 502560387
Encompass_Preferred
Chemical. CAS 1237-75-8. Formula C18H14BrNO3. MW BD9837. Naphthol AS-BI is a fluorescent dye commonly used as a building block or intermediate to produce fluorigenic substrates or as a coupling reagent to produce azo dyes. After cleavage of the fluorigenic substrates by esterases the free Naphthol AS-BI can couple with diazonium salts to form different types of fluorescent azo dyes.
Catalog No. 50-256-0387 Supplier AdipoGen Supplier No. CDXN0239G001
Chemical. CAS 1237-75-8. Formula C18H14BrNO3. MW BD9837. Naphthol AS-BI is a fluorescent dye commonly used as a building block or intermediate to produce fluorigenic substrates or as a coupling reagent to produce azo dyes. After cleavage of the fluorigenic substrates by esterases the free Naphthol AS-BI can couple with diazonium salts to form different types of fluorescent azo dyes.