Learn More
AdipoGen Naphthol AS-BI phosphate
SDP
Catalog No. 502560401
Encompass_Preferred
Chemical. CAS 1919-91-1. Formula C18H15BrNO6P. MW BD9837. Naphthol AS-BI-phosphate is a fluorogenic substrate for acid and alkaline phosphatases. Naphthol AS-BI-phosphate is converted to naphthol AS-BI N-ASBI displaying excitation/emission spectra of 405/515nm. N-ASBI fluorescence is a quantitative marker of acid and alkaline phosphatase activity. Acid and alkaline phosphatases are often used as clinical markers of disease, since the concentration of human acid and alkaline phosphatases undergo pronounced changes in particular diseases, resulting in unusually high or low concentrations. It is a histochemical substrate commonly used in conjunction with diazonium salts to form a specific azo dye for immunohistology, immunoblotting and dot blot applications.
Catalog No. 50-256-0401 Supplier AdipoGen Supplier No. CDXN0126G005
Chemical. CAS 1919-91-1. Formula C18H15BrNO6P. MW BD9837. Naphthol AS-BI-phosphate is a fluorogenic substrate for acid and alkaline phosphatases. Naphthol AS-BI-phosphate is converted to naphthol AS-BI N-ASBI displaying excitation/emission spectra of 405/515nm. N-ASBI fluorescence is a quantitative marker of acid and alkaline phosphatase activity. Acid and alkaline phosphatases are often used as clinical markers of disease, since the concentration of human acid and alkaline phosphatases undergo pronounced changes in particular diseases, resulting in unusually high or low concentrations. It is a histochemical substrate commonly used in conjunction with diazonium salts to form a specific azo dye for immunohistology, immunoblotting and dot blot applications.