Learn More
AdipoGen Naphthol AS-BI phosphate 2Na
SDP
Catalog No. 502560383
Encompass_Preferred
Chemical. CAS 207569-04-8. Formula C18H13BrNNa2O6P . xH2O. MW 496.16 anhydrous basis. Naphthol AS-BI-phosphate is a fluorogenic substrate for acid and alkaline phosphatases. Naphthol AS-BI-phosphate is converted to naphthol AS-BI N-ASBI displaying excitation/emission spectra of 405/515nm. N-ASBI fluorescence is a quantitative marker of acid and alkaline phosphatase activity. Acid and alkaline phosphatases are often used as clinical markers of disease, since the concentration of human acid and alkaline phosphatases undergo pronounced changes in particular diseases, resulting in unusually high or low concentrations.
Catalog No. 50-256-0383 Supplier AdipoGen Supplier No. CDXN0244G005
Chemical. CAS 207569-04-8. Formula C18H13BrNNa2O6P . xH2O. MW 496.16 anhydrous basis. Naphthol AS-BI-phosphate is a fluorogenic substrate for acid and alkaline phosphatases. Naphthol AS-BI-phosphate is converted to naphthol AS-BI N-ASBI displaying excitation/emission spectra of 405/515nm. N-ASBI fluorescence is a quantitative marker of acid and alkaline phosphatase activity. Acid and alkaline phosphatases are often used as clinical markers of disease, since the concentration of human acid and alkaline phosphatases undergo pronounced changes in particular diseases, resulting in unusually high or low concentrations.