Learn More
Cayman Chemical Naphthol AS-BI-Phosphate 500mg
SDP
Catalog No. 502985985
Encompass_Preferred
A fluorogenic substrate for acid and alkaline phosphatases; is converted to N-ASBI which displays ex/em spectra of 405/515 nm, respectively; fluorescence is a quantitative marker of acid and alkaline phosphatase activity
Catalog No. 50-298-5985 Supplier Cayman Chemical Supplier No. 21845500 MG
A fluorogenic substrate for acid and alkaline phosphatases; is converted to N-ASBI which displays ex/em spectra of 405/515 nm, respectively; fluorescence is a quantitative marker of acid and alkaline phosphatase activity