Learn More
Cayman Chemical Naphthol AS-BI-Phosphate 5g
SDP
Catalog No. 502986643
Encompass_Preferred
A fluorogenic substrate for acid and alkaline phosphatases; is converted to N-ASBI which displays ex/em spectra of 405/515 nm, respectively; fluorescence is a quantitative marker of acid and alkaline phosphatase activity
Catalog No. 50-298-6643 Supplier Cayman Chemical Supplier No. 218455 G
A fluorogenic substrate for acid and alkaline phosphatases; is converted to N-ASBI which displays ex/em spectra of 405/515 nm, respectively; fluorescence is a quantitative marker of acid and alkaline phosphatase activity