Learn More
Crescent Chemical Co Inc NAPHTHOL-AS-BI-PHOSPHATE
SDP
Supplier:  Crescent Chemical Co Inc SE29988.02
Encompass_Preferred
Naphthol-AS-BI-phosphate research grade Histochemical substrate for phosphatase. Size - 500MG Storage Conditions - -15 °C TO -25 °C Catalog Number - 29988.02 CAS 1919-91-1
Catalog No. 50-247-896