Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthol AS-BI Phosphate, Sigma-Aldrich

Catalog No. 01258817 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-817 5 g
01-258-896 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-817 Supplier MilliporeSigma Supelco Supplier No. N21255G
Add to Cart
Add to Cart

Naphthol AS-BI phosphate is suitable to use: in tartrate-resistant acid phosphatase (TRAP) staining to detect osteoclast activity as a component in Tris-HCl in staining cells to measure alkaline phosphatase (ALP) activity to perform alkaline phosphatase reaction.

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
Form Powder
Quantity 5 g
Substrate Properties Chemical Substrate
Substrate Type Naphthol Phosphate Substrate
Target Enzyme Alkaline Phosphatase
Product Type Substrates
Purity or Quality Grade Technical Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.