Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthol-AS-D chloroacetate, 98%, MP Biomedicals™

Catalog No. ICN10242401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10242401 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10242401 Supplier MP Biomedicals, Inc Supplier No. 0210242401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35245-26-2
Molecular Formula C20H16ClNO3
Molecular Weight (g/mol) 353.802
InChI Key FMVKYSCWHDVMGO-UHFFFAOYSA-N
Synonym naphthol as-d chloroacetate, 2-o-tolylcarbamoyl naphthalen-3-yl 2-chloroacetate, 3-hydroxy-2-naphthoic-o-toluidide chloroacetate, naphthol as-d-chloroacetate, 3-2-methylphenyl carbamoyl-2-naphthyl chloroacetate, 3-2-methylphenyl carbamoyl naphthalen-2-yl 2-chloroacetate, acetic acid, 2-chloro-,3-2-methylphenyl amino carbonyl-2-naphthalenyl ester, naphthol-as-d-chloroacetate, acetic acid, chloro-, s, naphthol as-d chloroacetate solution
PubChem CID 160247
IUPAC Name [3-[(2-methylphenyl)carbamoyl]naphthalen-2-yl] 2-chloroacetate
SMILES CC1=CC=CC=C1NC(=O)C2=CC3=CC=CC=C3C=C2OC(=O)CCl

Specifications

Color White
Quantity 1 g
Formula Weight 353.8
Percent Purity 0.98
Physical Form Powder
Chemical Name or Material Naphthol-AS-D chloroacetate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.