Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthol As-D Chloroacetate, MP Biomedicals™

Catalog No. MP021024241 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021024241 100 mg
MP021024245 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021024241 Supplier MP Biomedicals, Inc Supplier No. 02102424.1
Add to Cart
Add to Cart

Naphthol As-D Chloroacetate, MP Biomedicals is a substrate for histochemical localization of esterases.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35245-26-2
Molecular Formula C20H16ClNO3
Molecular Weight (g/mol) 353.802
InChI Key FMVKYSCWHDVMGO-UHFFFAOYSA-N
Synonym naphthol as-d chloroacetate, 2-o-tolylcarbamoyl naphthalen-3-yl 2-chloroacetate, 3-hydroxy-2-naphthoic-o-toluidide chloroacetate, naphthol as-d-chloroacetate, 3-2-methylphenyl carbamoyl-2-naphthyl chloroacetate, 3-2-methylphenyl carbamoyl naphthalen-2-yl 2-chloroacetate, acetic acid, 2-chloro-,3-2-methylphenyl amino carbonyl-2-naphthalenyl ester, naphthol-as-d-chloroacetate, acetic acid, chloro-, s, naphthol as-d chloroacetate solution
PubChem CID 160247
IUPAC Name [3-[(2-methylphenyl)carbamoyl]naphthalen-2-yl] 2-chloroacetate
SMILES CC1=CC=CC=C1NC(=O)C2=CC3=CC=CC=C3C=C2OC(=O)CCl

Specifications

Melting Point 161°C to 171°C
Quantity 100 mg
Solubility Information Soluble in ethyl acetate (5mg/mL, clear and slightly yellow solution); methanol (1mg/mL, clear and colorless solution) and acetone.
Formula Weight 353.8
Physical Form Powder
Recommended Storage Store at –20°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.