Learn More
Crescent Chemical Co Inc NAPHTHOL-AS-D-CHLOROACETATE
SDP
Supplier:  Crescent Chemical Co Inc SE29995.02
Encompass_Preferred
Naphthol-AS-D-chloroacetate pure Histochemical substrate for esterase with improved stability. Size - 1G Storage Conditions - -15 °C TO -25 °C Catalog Number - 29995.02 CAS 35245-26-2
Catalog No. 50-247-900