Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Naphthol AS-E | 92-78-4 | 98.0% | 297.74 | C17H12ClNO2 | 50 MG
SDP

Catalog No. 5000242508
Encompass_Preferred

Naphthol AS-E is a small-molecule, cell-permeable inhibitor of the KIX-KID protein-protein interaction used in biochemical and cell-based research. It binds the KIX domain of CBP and blocks the KIX-KID interaction, making it useful for mechanistic studies of transcriptional regulation.

  • Inhibits KIX-KID interaction with IC50 2.26 μM.
  • Binds CBP KIX domain (Kd ~8.6 μM).
  • High purity: 98.0%.
  • Molecular weight: 297.74 Da.
  • Soluble in DMSO (~41.67 mg/mL) for in vitro use.
  • Storage: powder stable at -20°C (up to 3 years) and 4°C (up to 2 years).
  • Available as a 50 MG solid for laboratory research.

Catalog No. 50-002-42508 Supplier Medchemexpress LLC Supplier No. HY10406850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Naphthol AS-E is a small-molecule, cell-permeable inhibitor of the KIX-KID protein-protein interaction used in biochemical and cell-based research. It binds the KIX domain of CBP and blocks the KIX-KID interaction, making it useful for mechanistic studies of transcriptional regulation.

  • Inhibits KIX-KID interaction with IC50 2.26 μM.
  • Binds CBP KIX domain (Kd ~8.6 μM).
  • High purity: 98.0%.
  • Molecular weight: 297.74 Da.
  • Soluble in DMSO (~41.67 mg/mL) for in vitro use.
  • Storage: powder stable at -20°C (up to 3 years) and 4°C (up to 2 years).
  • Available as a 50 MG solid for laboratory research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.