Learn More
Medchemexpress LLC Naphthol AS-E | 92-78-4 | MFCD00021639 | 98.0% | 297.74 | C17H12ClNO2 | 1 ML
SDP
Catalog No. 5000242561
Encompass_Preferred
Naphthol AS-E is a small-molecule inhibitor of the KIX-KID interaction that directly binds the KIX domain of CREB-binding protein (CBP). It is supplied as a solid or as a ready-to-use 10 mM solution in DMSO and is used in biochemical and cell-based assays to probe KIX-KID-mediated interactions.
  • Potent, cell-permeable inhibitor of the KIX-KID interaction.
  • Directly binds the KIX domain of CBP (reported Kd ≈ 8.6 μM).
  • Available as solid or 10 mM solution in DMSO for ready use.
  • High purity suitable for research applications.
  • Stable under recommended storage conditions for powder and in solvent.
Catalog No. 50-002-42561 Supplier Medchemexpress LLC Supplier No. HY10406810MM 1ML
Naphthol AS-E is a small-molecule inhibitor of the KIX-KID interaction that directly binds the KIX domain of CREB-binding protein (CBP). It is supplied as a solid or as a ready-to-use 10 mM solution in DMSO and is used in biochemical and cell-based assays to probe KIX-KID-mediated interactions.
  • Potent, cell-permeable inhibitor of the KIX-KID interaction.
  • Directly binds the KIX domain of CBP (reported Kd ≈ 8.6 μM).
  • Available as solid or 10 mM solution in DMSO for ready use.
  • High purity suitable for research applications.
  • Stable under recommended storage conditions for powder and in solvent.