Learn More
Alkali Scientific Naphthol AS MX phosphate, 100 Mg
SDP
Catalog No. 5000660624
Encompass_Preferred
Naphthol AS MX Phosphate (APH002 CP) is a reagent used in laboratory experiments involving enzyme activity assays. This 100mg package is specifically used as a phosphate substrate for enzyme reactions, particularly in histochemistry and cytochemistry studies. The naphthol-based substrate undergoes a colorimetric change when enzymatically cleaved, providing a visible indication of enzyme activity. Naphthol AS MX Phosphate is used to study the activity of phosphatases and other enzymes in cellular and tissue samples. This compound is often employed in diagnostic and research applications, particularly in the study of enzyme-linked immunosorbent assays (ELISA) and other biochemical tests.
Catalog No. 50-006-60624 Supplier Alkali Scientific Supplier No. APH002CP
Naphthol AS MX Phosphate (APH002 CP) is a reagent used in laboratory experiments involving enzyme activity assays. This 100mg package is specifically used as a phosphate substrate for enzyme reactions, particularly in histochemistry and cytochemistry studies. The naphthol-based substrate undergoes a colorimetric change when enzymatically cleaved, providing a visible indication of enzyme activity. Naphthol AS MX Phosphate is used to study the activity of phosphatases and other enzymes in cellular and tissue samples. This compound is often employed in diagnostic and research applications, particularly in the study of enzyme-linked immunosorbent assays (ELISA) and other biochemical tests.