Learn More
Medchemexpress LLC Naphthol AS-MX phosphate | 1596-56-1 | MFCD00042711 | 99.5% | 371.32 g/mol | C19H18NO5P | 50 MG
SDP
Catalog No. 5000243088
Encompass_Preferred
Naphthol AS-MX phosphate is the phosphate salt used as a chromogenic and fluorogenic substrate for phosphatase-based assays and histochemistry. Supplied as a high-purity solid, it is suitable for enzymatic activity assays, immunoblotting, and histochemical staining; store sealed and protected from moisture, with recommended low-temperature storage for solutions.
Catalog No. 50-002-43088 Supplier Medchemexpress LLC Supplier No. HY10406750MG
Naphthol AS-MX phosphate is the phosphate salt used as a chromogenic and fluorogenic substrate for phosphatase-based assays and histochemistry. Supplied as a high-purity solid, it is suitable for enzymatic activity assays, immunoblotting, and histochemical staining; store sealed and protected from moisture, with recommended low-temperature storage for solutions.