Learn More
AdipoGen Naphthol AS-MX phosphate 2Na
SDP
Catalog No. 502560379
Encompass_Preferred
Chemical. CAS 96189-12-7. Formula C19H16NO5PNa2. MW BD9837. Naphthol AS-MX Phosphate is a fluorogenic substrate for alkaline phosphatases. Naphthol AS-MX Phosphate is converted to naphthol AS-MX displaying excitation/emission spectra of 388/512nm. The fluorescence is a quantitative marker of acid and alkaline phosphatase activity. Acid and alkaline phosphatases are often used as clinical markers of disease, since the concentration of human acid and alkaline phosphatases undergo pronounced changes in particular diseases, resulting in unusually high or low concentrations. It is a histochemical substrate commonly used in conjunction with diazonium salts to form a specific azo dye for immunohistology, immunoblotting and dot blot applications.
Catalog No. 50-256-0379 Supplier AdipoGen Supplier No. CDXN0246M500
Chemical. CAS 96189-12-7. Formula C19H16NO5PNa2. MW BD9837. Naphthol AS-MX Phosphate is a fluorogenic substrate for alkaline phosphatases. Naphthol AS-MX Phosphate is converted to naphthol AS-MX displaying excitation/emission spectra of 388/512nm. The fluorescence is a quantitative marker of acid and alkaline phosphatase activity. Acid and alkaline phosphatases are often used as clinical markers of disease, since the concentration of human acid and alkaline phosphatases undergo pronounced changes in particular diseases, resulting in unusually high or low concentrations. It is a histochemical substrate commonly used in conjunction with diazonium salts to form a specific azo dye for immunohistology, immunoblotting and dot blot applications.