Learn More
Crescent Chemical Co Inc NAPHTHOL-AS-MX-PHOSPHATE
SDP
Catalog No. 50247901
Encompass_Preferred
Naphthol-AS-MX-phosphate research grade Selective substrate of alkaline phosphatase. Suitable for dye coupling. Fast Blue RR (1). Fast Red TR-salt (cat. no. 21317) (2). Size - 250MG Storage Conditions - -15 °C TO -25 °C Catalog Number - 30002.01 CAS No.
Catalog No. 50-247-901 Supplier Crescent Chemical Co Inc Supplier No. SE30002.01
Naphthol-AS-MX-phosphate research grade Selective substrate of alkaline phosphatase. Suitable for dye coupling. Fast Blue RR (1). Fast Red TR-salt (cat. no. 21317) (2). Size - 250MG Storage Conditions - -15 °C TO -25 °C Catalog Number - 30002.01 CAS No.