Learn More
Alkali Scientific Naphthol AS MX phosphate, 5 Grams
SDP
Supplier:  Alkali Scientific APH001CP
Encompass_Preferred
Naphthol AS MX Phosphate is an enzyme substrate used in laboratory applications to detect and quantify phosphatase activity. It is commonly used in enzyme assays where naphthol AS MX phosphate undergoes hydrolysis by phosphatases, resulting in a colored product. This product is widely employed in immunohistochemistry, enzyme-linked immunosorbent assays (ELISA), and various other biochemical studies. The 5g quantity is ideal for small-scale experiments, providing enough material for multiple assays. It is used to investigate enzyme kinetics, study phosphatase inhibitors, and explore cellular signaling mechanisms. This reagent is essential in laboratories studying enzyme function and related biochemical pathways.
Catalog No. 50-006-61539