Learn More
Sigma Aldrich Fine Chemicals Biosciences Naphthol AS-MX phosphate p1G
Catalog No. 502902214
Encompass
Naphthol AS-MX phosphate is soluble in dioxane and is a substrate for phosphatase.
Catalog No. 50-290-2214 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N48751G
Naphthol AS-MX phosphate is soluble in dioxane and is a substrate for phosphatase.