Learn More
AdipoGen Naphthol AS phosphate 2Na
SDP
Catalog No. 502560385
Encompass_Preferred
Chemical. CAS 69815-54-9. Formula C17H12NNa2O5P. MW BD9837. Naphthol AS phosphate is a fluorogenic substrate for acid and alkaline phosphatases. Naphthol AS phosphate is converted to naphthol AS displaying excitation/emission spectra of 388/516nm. The fluorescence is a quantitative marker of acid and alkaline phosphatase activity. Acid and alkaline phosphatases are often used as clinical markers of disease, since the concentration of human acid and alkaline phosphatases undergo pronounced changes in particular diseases, resulting in unusually high or low concentrations. It is a histochemical substrate commonly used in conjunction with diazonium salts to form a specific azo dye for immunohistology, immunoblotting, dot blot applications and in situ PCR.
Catalog No. 50-256-0385 Supplier AdipoGen Supplier No. CDXN0240G005
Chemical. CAS 69815-54-9. Formula C17H12NNa2O5P. MW BD9837. Naphthol AS phosphate is a fluorogenic substrate for acid and alkaline phosphatases. Naphthol AS phosphate is converted to naphthol AS displaying excitation/emission spectra of 388/516nm. The fluorescence is a quantitative marker of acid and alkaline phosphatase activity. Acid and alkaline phosphatases are often used as clinical markers of disease, since the concentration of human acid and alkaline phosphatases undergo pronounced changes in particular diseases, resulting in unusually high or low concentrations. It is a histochemical substrate commonly used in conjunction with diazonium salts to form a specific azo dye for immunohistology, immunoblotting, dot blot applications and in situ PCR.