Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthol AS Phosphate Disodium Salt, Sigma-Aldrich

Catalog No. 01258890 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-890 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-890 Supplier MilliporeSigma Supelco Supplier No. N92525G
Add to Cart
Add to Cart

Histochemical substrate for acid and alkaline phosphatase.

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Colorimetric
Form Powder
Quantity 5 g
Substrate Type Naphthol Phosphate Substrate
Target Enzyme Alkaline Phosphatase
Product Type Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.