Learn More
Medchemexpress LLC Naphthol AS-TR phosphate disodium | 4264-93-1 | MFCD00069550 | 99.9% | C18H13ClNNa2O5P | 10MG
SDP
Supplier:  Medchemexpress LLC HY130767A10MG
Encompass_Preferred
Naphthol AS-TR phosphate disodium is a water-soluble enzymatic substrate used to detect alkaline phosphatase activity. The disodium salt is hydrolyzed by phosphatases to form a colored product that is measurable spectrophotometrically, making it suitable for biochemical assays, histochemical staining, and enzyme activity monitoring in biological samples.
  • Water-soluble disodium salt suitable for aqueous assays.
  • Sensitive substrate for alkaline and acid phosphatases.
  • Generates a colored product upon enzymatic hydrolysis for spectrophotometric detection.
  • High purity for reproducible assay results.
  • Available in small pack sizes for laboratory-scale use.
  • Stable solid form for storage and handling.
Catalog No. 50-002-59627