Learn More
Medchemexpress LLC Naphthol green B 100g
SDP
Catalog No. 5000206551
Encompass_Preferred
Naphthol green B is an iron-complex dye Naphthol green B can be used in wool dyeing silk dyeing and polyamide dyeing Naphthol green B can be used to fabricate electrochemical sensor for the detection of H2O2[1][2]
Catalog No. 50-002-06551 Supplier Medchemexpress LLC Supplier No. HYW110905100G
Naphthol green B is an iron-complex dye Naphthol green B can be used in wool dyeing silk dyeing and polyamide dyeing Naphthol green B can be used to fabricate electrochemical sensor for the detection of H2O2[1][2]