Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthol Green B Dye, 2.5% w/v, Reagents

Catalog No. 01334680
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-680 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-680 Supplier Reagents Holdings Llc Supplier No. CN00250010F
Add to Cart
Add to Cart

Naphthol Green B Dye, 2.5% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Naphthol Green B Dye, 2.5% w/v
CAS 7732-18-5, 19381-50-1
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.01
Color Dark Green
Quantity 10 L
Physical Form Liquid
Percent Purity 2.5%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.