Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthol Yellow S, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015675 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5675 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5675 Supplier MilliporeSigma Supelco Supplier No. 4954725MG
Add to Cart
Add to Cart

Naphthol Yellow S is a dye that is known for its hazardous nature.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 846-70-8
Physical Form Neat
Linear Formula C10H4N2 Na2O8 S
MDL Number MFCD00003958
Synonym 2,4-Dinitro-1-naphthol-7-sulfonic acid sodium salt; 5,7-Dinitro-8-hydroxy-2-naphthalenesulfonic acid sodium salt; Acid Yellow 1; Flavianic acid sodium salt
Type Analytical Standard
Quantity 25 mg
Formula Weight 358.19
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.