Learn More
AdipoGen Naphtholfluorescein diacetate
SDP
Catalog No. 502560435
Encompass_Preferred
Chemical. CAS 77084-71-0. Formula C32H20O7. Molecular Weight 516.5. Naphthofluorescein-diacetate is a non-fluoresent compound that is hydrolyzed by esterases to the fluorescent compound Naphthofluorescein. Spectral data lambdaex 594 nm lambdaem 663 nm in 0.1 M Tris pH 9.0 esterase. The fluorescent naphthofluorescein has been shown to be an inhibitor of furin.
Catalog No. 50-256-0435 Supplier AdipoGen Supplier No. CDXN0028M100
Chemical. CAS 77084-71-0. Formula C32H20O7. Molecular Weight 516.5. Naphthofluorescein-diacetate is a non-fluoresent compound that is hydrolyzed by esterases to the fluorescent compound Naphthofluorescein. Spectral data lambdaex 594 nm lambdaem 663 nm in 0.1 M Tris pH 9.0 esterase. The fluorescent naphthofluorescein has been shown to be an inhibitor of furin.