Learn More
AdipoGen Naphthomycin B
SDP
Catalog No. 502562024
Encompass_Preferred
Chemical. CAS 86825-88-9. Formula C39H44ClNO9. MW 706.2. Isolated from Streptomyces sp. Gö 40/14. Ansamycin antibiotic. Antibacterial and antifungal.
Catalog No. 50-256-2024 Supplier AdipoGen Supplier No. BVT0265M001
Chemical. CAS 86825-88-9. Formula C39H44ClNO9. MW 706.2. Isolated from Streptomyces sp. Gö 40/14. Ansamycin antibiotic. Antibacterial and antifungal.