Learn More
Medchemexpress LLC Naphthoquine phosphate | 173531-58-3 | 99.6% | 605.94 | 100 MG
SDP
Catalog No. 5000384236
Encompass_Preferred
Naphthoquine phosphate is a potent, orally active antimalarial agent with a thorough killing function for various schizonts of plasmodia, including those resistant to Chloroquine. In vivo studies have shown a parasite reduction rate higher than 90% in treated monkeys. This product is for research use only.
  • Potent and orally active antimalarial agent
  • Exhibits thorough killing function against various schizonts of plasmodia
  • Effective against P. falciparum resistant to Chloroquine
  • Demonstrates high parasite reduction rates in vivo
Catalog No. 50-003-84236 Supplier Medchemexpress LLC Supplier No. HY17036100MG
Naphthoquine phosphate is a potent, orally active antimalarial agent with a thorough killing function for various schizonts of plasmodia, including those resistant to Chloroquine. In vivo studies have shown a parasite reduction rate higher than 90% in treated monkeys. This product is for research use only.
  • Potent and orally active antimalarial agent
  • Exhibits thorough killing function against various schizonts of plasmodia
  • Effective against P. falciparum resistant to Chloroquine
  • Demonstrates high parasite reduction rates in vivo