Learn More
Medchemexpress LLC Naphthoquine phosphate | 173531-58-3 | MFCD25541748 | 99.6% | 605.94 g/mol | C24H34ClN3O9P2 | 10 MG
SDP
Catalog No. 5000295105
Encompass_Preferred
Naphthoquine phosphate is an orally active antimalarial research compound that shows potent activity against Plasmodium schizonts and rapid parasite reduction in animal models. Supplied as a research-grade solid, the product includes manufacturer guidance for solubility and storage to support laboratory formulation and dosing studies.
  • Orally active antimalarial with schizonticidal activity.
  • Demonstrates rapid parasite reduction in in vivo models.
  • Supplied as a solid suitable for formulation and dosing studies.
  • Water-soluble at measured concentration; limited solubility in DMSO.
  • Recommended sealed storage at 4°C; solvent stability data provided for -80°C and -20°C.
  • For research use only; not intended for human therapeutic use.
Catalog No. 50-002-95105 Supplier Medchemexpress LLC Supplier No. HY1703610MG
Naphthoquine phosphate is an orally active antimalarial research compound that shows potent activity against Plasmodium schizonts and rapid parasite reduction in animal models. Supplied as a research-grade solid, the product includes manufacturer guidance for solubility and storage to support laboratory formulation and dosing studies.
  • Orally active antimalarial with schizonticidal activity.
  • Demonstrates rapid parasite reduction in in vivo models.
  • Supplied as a solid suitable for formulation and dosing studies.
  • Water-soluble at measured concentration; limited solubility in DMSO.
  • Recommended sealed storage at 4°C; solvent stability data provided for -80°C and -20°C.
  • For research use only; not intended for human therapeutic use.