Learn More
Crescent Chemical Co Inc NAPHTHYL ACETATE ANALYTICAL
SDP
Catalog No. 50247902
Encompass_Preferred
Naphthyl acetate analytical grade Substrate for esterases. Size - 25G Storage Conditions - +2 °C TO +8 °C Catalog Number - 30040.01 CAS 830-81-9
Catalog No. 50-247-902 Supplier Crescent Chemical Co Inc Supplier No. SE30040.01
Naphthyl acetate analytical grade Substrate for esterases. Size - 25G Storage Conditions - +2 °C TO +8 °C Catalog Number - 30040.01 CAS 830-81-9