Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Naphthyl Laurate, MP Biomedicals

Catalog No. ICN221195 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN221195 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN221195 Supplier MP Biomedicals, Inc Supplier No. 0522119510

Chemical Identifiers

CAS 4227-99-0
Molecular Formula C22H30O2
Molecular Weight (g/mol) 326.48
InChI Key CJSQRFDKBOSEJH-UHFFFAOYSA-N
Synonym alpha-naphthyl laurate, dodecanoic acid 1-naphthyl ester, 1-naphthyllaurate, naphthyl dodecanoate, 1-naphthyl laurate, 1-naphthyl dodecanoate, 1-naphthalenyl dodecanoate, lauric acid 1-naphthyl ester
PubChem CID 4112759
IUPAC Name naphthalen-1-yl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC1=CC=CC2=CC=CC=C21

Specifications

Quantity 10 g
Formula Weight 326.48
Chemical Name or Material Alpha-Naphthyl Laurate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.