Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Naphthyl Phosphate Disodium Salt Hydrate , MP Biomedical

Catalog No. MP021002571 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021002571 100 mg
MP021002572 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021002571 Supplier MP Biomedicals, Inc Supplier No. 02100257.1
Add to Cart
Add to Cart

α-Naphthyl Phosphate Disodium Salt Hydrate is a white, crystalline powder enzyme substrate used for cell biology and analysis.

  • Substrate for determination of phosphatase
  • Used as an organic chemical synthesis intermediate
TRUSTED_SUSTAINABILITY

Specifications

Color White
Quantity 100 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.