Learn More
Naphthylethylenediamine Dihydrochloride, 0.1% (w/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Color | Brown to Green-Yellow |
| CAS | 1465-25-4, 7732-18-5 |
| CAS Min % | 98.0, 0.1 |
| CAS Max % | 100.0, 0.1 |
| Synonym | pentachlorobenzene, benzene, pentachloro, rcra waste number u183, unii-d62gwo6832, ccris 5927, qcb, rcra waste no. u183, 1,2,3,4,5-pentachloro-benzene, benzene, 1,2,3,4,5-pentachloro, pentachlorbenzol |
| SMILES | [H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1 |
| IUPAC Name | dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride |
| Molecular Weight (g/mol) | 259.17 |
| PubChem CID | 11855 |
| ChEBI | CHEBI:47136 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.