Learn More
Cayman Chemical Naphyrone 1-naphthyl isomer (hydrochloride), Purity: ≥95%, CAS: 1349245-31-3, 1 mg
SDP
Catalog No. 502951234
Encompass_Preferred
A structural isomer of naphyrone, having the napthyl group attached at the 1, rather than 2, position; intended for forensic and research applications
Catalog No. 50-295-1234 Supplier Cayman Chemical Supplier No. 112401 MG
A structural isomer of naphyrone, having the napthyl group attached at the 1, rather than 2, position; intended for forensic and research applications