Learn More
Cayman Chemical Naphyron 1naphthyl Isomer 10mg
SDP
Catalog No. 502951409
Encompass_Preferred
A structural isomer of naphyrone, having the napthyl group attached at the 1, rather than 2, position; intended for forensic and research applications
Catalog No. 50-295-1409 Supplier Cayman Chemical Supplier No. 1124010 MG
A structural isomer of naphyrone, having the napthyl group attached at the 1, rather than 2, position; intended for forensic and research applications