Learn More
Selleck Chemical LLC Naporafenib S8745-25MG
SDP
Catalog No. 5000024446
Encompass_Preferred
Naporafenib (LXH254) is a type II ATP-competitive inhibitor that inhibits both B- and CRAF kinase activities at picomolar concentrations with a high degree of selectivity against a panel of 456 human kinases and in cell-based assays
Catalog No. 50-000-24446 Supplier Selleck Chemical LLC Supplier No. S874525MG
Naporafenib (LXH254) is a type II ATP-competitive inhibitor that inhibits both B- and CRAF kinase activities at picomolar concentrations with a high degree of selectivity against a panel of 456 human kinases and in cell-based assays