Learn More
Medchemexpress LLC Napropamide | 15299-99-7 | MFCD00055354 | 99.4% | 271.35 | C17H21NO2 | 10 MG
SDP
Catalog No. 5000296806
Encompass_Preferred
Analytical reference standard of napropamide supplied as a 10 mg material for research and analytical applications. The compound is provided with high purity to support qualitative and quantitative assays and method development in chromatography and mass spectrometry.
  • Analytical reference standard suitable for assays and method validation.
  • High purity (99.39%), supporting accurate quantitative results.
  • Small 10 mg quantity for analytical workflows and stability testing.
  • Suitable for HPLC, GC, and MS applications.
  • Certificate of analysis lists recommended storage conditions.
Catalog No. 50-002-96806 Supplier Medchemexpress LLC Supplier No. HYB1972R10MG
Analytical reference standard of napropamide supplied as a 10 mg material for research and analytical applications. The compound is provided with high purity to support qualitative and quantitative assays and method development in chromatography and mass spectrometry.
  • Analytical reference standard suitable for assays and method validation.
  • High purity (99.39%), supporting accurate quantitative results.
  • Small 10 mg quantity for analytical workflows and stability testing.
  • Suitable for HPLC, GC, and MS applications.
  • Certificate of analysis lists recommended storage conditions.