Learn More
Medchemexpress LLC Napropamide (Standard) | 15299-99-7 | 99.4% | 500 MG
SDP
Catalog No. 5000410877
Encompass_Preferred
Napropamide (Standard) is the analytical standard of Napropamide, an amide herbicide used to control weeds in fruits, vegetables, tobacco, and ornamental plants. It primarily inhibits DNA synthesis and cell division and exhibits moderate to high persistence in the field, with a half-life of 24-131 days. This product is susceptible to photodegradation.
  • Intended for research and analytical applications.
  • Grade of analytical standard and reference standard supplied assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-10877 Supplier Medchemexpress LLC Supplier No. HYB1972500MG
Napropamide (Standard) is the analytical standard of Napropamide, an amide herbicide used to control weeds in fruits, vegetables, tobacco, and ornamental plants. It primarily inhibits DNA synthesis and cell division and exhibits moderate to high persistence in the field, with a half-life of 24-131 days. This product is susceptible to photodegradation.
  • Intended for research and analytical applications.
  • Grade of analytical standard and reference standard supplied assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.