Learn More
Selleck Chemical LLC Naproxen S5177-25mg
SDP
Catalog No. 503108253
Encompass_Preferred
Naproxen is an anti-inflammatory agent with analgesic and antipyretic properties It is a nonselective inhibitor of COX-1 and COX-2
Catalog No. 50-310-8253 Supplier Selleck Chemical LLC Supplier No. S517725MG
Naproxen is an anti-inflammatory agent with analgesic and antipyretic properties It is a nonselective inhibitor of COX-1 and COX-2