Learn More
Cayman Chemical -Naproxen-d3 1mg
SDP
Catalog No. 502980368
Encompass_Preferred
An internal standard for the quantification of naproxen by GC- or LC-MS
Catalog No. 50-298-0368 Supplier Cayman Chemical Supplier No. 182581 MG
An internal standard for the quantification of naproxen by GC- or LC-MS