Learn More
Medchemexpress LLC (±)-Naproxen-d3 | 958293-79-3 | C14H11D3O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY15029S1MG
Encompass_Preferred
(±)-Naproxen-d3 is the deuterium labeled (±)-Naproxen, which is a non-steroidal anti-inflammatory drug (NSAID).
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-75319