Learn More
Medchemexpress LLC (±)-Naproxen-d3 | 958293-79-3 | C14H11D3O3 | 5 MG

Supplier: Medchemexpress LLC HY15029S5MG
(±)-Naproxen-d3 is the deuterium labeled (±)-Naproxen, which is a non-steroidal anti-inflammatory drug (NSAID). It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
- Purity of 99.77%
- Can be used as a tracer
- Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
- Molecular weight of 233.28
- Solid appearance
- Off-white to light yellow color
- Soluble in DMSO at 100 mg/mL
- Powder stable for 3 years at -20°C and 2 years at 4°C
- Stable in solvent for 6 months at -80°C and 1 month at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.