Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (±)-Naproxen-d3 | 958293-79-3 | MFCD22572922 | 99.8% | 233.28 g/mol | C14H11D3O3 | 10 MG
SDP

Catalog No. 5000286387
Encompass_Preferred

(±)-Naproxen-d3 is the deuterium-labeled form of naproxen used as an analytical reference standard and tracer. It is intended as an internal standard for quantitative and metabolic studies and supports accurate method development and validation in spectrometric and chromatographic workflows.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High chemical and isotopic purity for reliable quantitation.
  • Supplied in small, accurately weighed quantities for analytical work.
  • Molecular formula C14H11D3O3 and molecular weight 233.28 g/mol.

Catalog No. 50-002-86387 Supplier Medchemexpress LLC Supplier No. HY15029S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(±)-Naproxen-d3 is the deuterium-labeled form of naproxen used as an analytical reference standard and tracer. It is intended as an internal standard for quantitative and metabolic studies and supports accurate method development and validation in spectrometric and chromatographic workflows.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High chemical and isotopic purity for reliable quantitation.
  • Supplied in small, accurately weighed quantities for analytical work.
  • Molecular formula C14H11D3O3 and molecular weight 233.28 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.