Learn More
Medchemexpress LLC Naproxen etemesil | 385800-16-8 | 99.89% | 336.40 | 50 MG
SDP
Catalog No. 5000401306
Encompass_Preferred
Naproxen etemesil is a lipophilic, non-acidic, inactive proagent of naproxen that is hydrolyzed to pharmacologically active Naproxen once absorbed. Naproxen is a COX-1 and COX-2 inhibitor with IC50s of 8.72 and 5.15 μM, respectively, in cell assay. It is for research use only.
  • Lipophilic, non-acidic, inactive prodrug of naproxen
  • Hydrolyzed to pharmacologically active Naproxen upon absorption
  • Inhibits COX-1 and COX-2 with IC50s of 8.72 μM and 5.15 μM, respectively, in cell assay
  • Exerts anti-inflammatory and antifibrotic effects in a mouse model of bleomycin-induced lung fibrosis
  • Downregulates TGF-β levels and Smad3/4 complex formation
  • Inhibits inflammatory pain, fever, and PGE2 synthesis
Catalog No. 50-004-01306 Supplier Medchemexpress LLC Supplier No. HY1967550MG
Naproxen etemesil is a lipophilic, non-acidic, inactive proagent of naproxen that is hydrolyzed to pharmacologically active Naproxen once absorbed. Naproxen is a COX-1 and COX-2 inhibitor with IC50s of 8.72 and 5.15 μM, respectively, in cell assay. It is for research use only.
  • Lipophilic, non-acidic, inactive prodrug of naproxen
  • Hydrolyzed to pharmacologically active Naproxen upon absorption
  • Inhibits COX-1 and COX-2 with IC50s of 8.72 μM and 5.15 μM, respectively, in cell assay
  • Exerts anti-inflammatory and antifibrotic effects in a mouse model of bleomycin-induced lung fibrosis
  • Downregulates TGF-β levels and Smad3/4 complex formation
  • Inhibits inflammatory pain, fever, and PGE2 synthesis