Learn More
Medchemexpress LLC Naproxen glucuronide 1mg
SDP
Catalog No. 5000205068
Encompass_Preferred
Naproxen glucuronide ((S)-Naproxen- -D-glucuronide) is a non-selective COX inhibitor Naproxen glucuronide a metabolite of naproxen is a nonsteroidal anti-inflammatory drug (NSAID) of the propionic acid class (the same as ibuprofen) that relieves pain fever swelling and stiffness[1]
Catalog No. 50-002-05068 Supplier Medchemexpress LLC Supplier No. HYW1219011MG
Naproxen glucuronide ((S)-Naproxen- -D-glucuronide) is a non-selective COX inhibitor Naproxen glucuronide a metabolite of naproxen is a nonsteroidal anti-inflammatory drug (NSAID) of the propionic acid class (the same as ibuprofen) that relieves pain fever swelling and stiffness[1]