Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naproxen Sodium, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18604748 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-748 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-604-748 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N1026100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N1026, 26159-34-2, C14H13NaO3

Spectrum™ Chemical Naproxen Sodium, USP is a non-steroidal anti-inflammatory drug. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 26159-34-2
Molecular Formula C14H13NaO3
Molecular Weight (g/mol) 252.25
MDL Number MFCD00058507
InChI Key CDBRNDSHEYLDJV-FVGYRXGTSA-M
IUPAC Name sodium (2S)-2-(6-methoxynaphthalen-2-yl)propanoate
SMILES [Na+].COC1=CC=C2C=C(C=CC2=C1)[C@H](C)C([O-])=O

Specifications

CAS Max % 1
Loss on Drying 0.01
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation -15.3° to -17.0°
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.