Learn More
AdipoGen Naproxen Solution (5 ml)
SDP
Catalog No. 502054562
Encompass_Preferred
Chemical. CAS: 22204-53-1. Formula: C14H14O3. MW: 230.26. Naproxen is a potent, non-steroidal, non-selective anti-inflammatory cyclooxygenase (COX-1 and COX-2) inhibitor. Inhibits COX-1 expression in HUVECS at a concentration of 5µg/ml. This COX inhibitor is indicated for the treatment of pain, fever, inflammation and stiffness. Research indicates that the S-naproxen is a more active form than the R-naproxen. The S-naproxen enantiomer was observed to inhibit platelet aggregation, and both enantiomers decrease production of thromboxane B2. Naproxen has been demonstrated to block A? fibril growth and form a complex with copper (II).
Catalog No. 50-205-4562 Supplier AdipoGen Supplier No. CDXN0258L005
Chemical. CAS: 22204-53-1. Formula: C14H14O3. MW: 230.26. Naproxen is a potent, non-steroidal, non-selective anti-inflammatory cyclooxygenase (COX-1 and COX-2) inhibitor. Inhibits COX-1 expression in HUVECS at a concentration of 5µg/ml. This COX inhibitor is indicated for the treatment of pain, fever, inflammation and stiffness. Research indicates that the S-naproxen is a more active form than the R-naproxen. The S-naproxen enantiomer was observed to inhibit platelet aggregation, and both enantiomers decrease production of thromboxane B2. Naproxen has been demonstrated to block A? fibril growth and form a complex with copper (II).